Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotang Inc BioAcryl-P (30%) Bis-Acrylamide 19:1, 1000 ml

Catalog No. 50151944 Shop All BIOTANG Inc Products
Encompass_Preferred

BioAcryl-P (30%) Bis-Acrylamide 19:1, 1000 ml.

Catalog No. 50-151-944 Supplier Biotang Inc Supplier No. BTBB277
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BioAcryl-P (30%) Bis-Acrylamide 19:1, 1000 ml.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.