Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioAcryl-P (40%, 29:1)

Catalog No. AAJ63079K2
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63079K2 1 L
AAJ63079AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63079K2 Supplier Thermo Scientific Chemicals Supplier No. J63079K2
Add to Cart

missing translation for 'validMainframeMsg'

BioAcryl-P (Acrylamide/Bisacrylamide 29:1, CAS # 25034-58-6) is a powder-based formula used to create the gel matrix necessary for electrophoresis experiments.

Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • BioAcryl-P is a 29:1 ratio of Acrylamide/Bisacrylamide
  • Acrylamide is a crystalline amide that forms polymer structures at room temperature
  • Acrylamide acts as the monomer for the gel structure, and bisacrylamide as the crosslinker for gel matrix formation

Application

  • Suitable for gel electrophoresis-mediated separation of proteins and DNA for identification or sequencing
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 29:1
Physical Form Liquid
Concentration 40% soln
Color Colorless
Quantity 1 L
MDL Number MFCD00080848
UN Number UN3426
Solubility Information It is soluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.