Learn More
Apexbio Technology LLC DiscoveryProbe™ Bioactive Compound Library Plus, 250uL/well(10 mM solution), 96 Well Racks With Screw Cap
SDP
Catalog No. 501998303
Encompass_Preferred
DiscoveryProbe™ Bioactive Compound Library Plus is a ready-to-use chemical library used for drug discovery, lab drug screening, drug target identification, and other pharmaceutical-related applications. It is available for high-throughput screening (HTS) and high-content screening (HCS). It consists of over 4400 small molecules with validated biological and pharmacological activities. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8303 Supplier Apexbio Technology LLC Supplier No. L1022P.25
Add to Cart
Add to Cart
Request a Quote
DiscoveryProbe™ Bioactive Compound Library Plus is a ready-to-use chemical library used for drug discovery, lab drug screening, drug target identification, and other pharmaceutical-related applications. It is available for high-throughput screening (HTS) and high-content screening (HCS). It consists of over 4400 small molecules with validated biological and pharmacological activities. Other sizes are also available. Please inqury us for quote.