Learn More
BioVendor, LLC Human Menopausal Gonadotropin (hMG), Native Protein (10 mg)
SDP
Catalog No. 502069284
Encompass
Human Menopausal Gonadotropin (hMG), Native Protein (10 mg). Research use only. hMG is produced from a sterile preparation of placental glucoprotein urine of post-menopausal women. Urine of postmenopausal women reflects the hypergonadotropic state of menopause: levels of follicle stimulating hormone (FSH) and luteinizing hormone (LH) are high. The MG is purified by proprietary chromatographic techniques. Appearance: Sterile Filtered White lyophilized (freeze-dried) powder. Formulation: The Human MG was lyophilized from a concentrated (1 mg/ml) solution with no additives. Biological activity: 100 IU/mg of FSH and 100 IU/mg LH. Synonyms: Menotropin.
Catalog No. 50-206-9284 Supplier BioVendor, LLC Supplier No. RP178251010
Human Menopausal Gonadotropin (hMG), Native Protein (10 mg). Research use only. hMG is produced from a sterile preparation of placental glucoprotein urine of post-menopausal women. Urine of postmenopausal women reflects the hypergonadotropic state of menopause: levels of follicle stimulating hormone (FSH) and luteinizing hormone (LH) are high. The MG is purified by proprietary chromatographic techniques. Appearance: Sterile Filtered White lyophilized (freeze-dried) powder. Formulation: The Human MG was lyophilized from a concentrated (1 mg/ml) solution with no additives. Biological activity: 100 IU/mg of FSH and 100 IU/mg LH. Synonyms: Menotropin.