Learn More
Enzo Life Sciences SCREEN-WELL Bioactive lipid library (500 µl/well)
SDP
Catalog No. 502010178
Encompass_Preferred
The SCREEN-WELL Bioactive Lipid Library contains 190 bioactive lipids in a 96 well format. Ideal for screening or identifiying recombinant orphan G protein-coupled receptors, target validation, secondary screening, validating new assays and for routine pharmacological applications. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0178 Supplier Enzo Life Sciences Supplier No. BML28000500
Add to Cart
Add to Cart
Request a Quote
The SCREEN-WELL Bioactive Lipid Library contains 190 bioactive lipids in a 96 well format. Ideal for screening or identifiying recombinant orphan G protein-coupled receptors, target validation, secondary screening, validating new assays and for routine pharmacological applications. Long Term Storage: -80°C. Applications: HTS .