Learn More
Gyros Inc BIOAFFY 1000
SDP
Catalog No. NC9021271
Encompass
one (1) Gyrolab Bioaffy CD microlaboratory, 1000 nL sample volume, 96 microstructures.
This item is not returnable. View return policy
Catalog No. NC9021271 Supplier Gyros Inc Supplier No. P0004253
one (1) Gyrolab Bioaffy CD microlaboratory, 1000 nL sample volume, 96 microstructures.