Learn More
Agilent Technologies Agilent DNA 7500 Kit
Catalog No. NC1762600
Encompass
For the separation, sizing and quantification of dsDNA fragments from 100 to 7500 bp. The kit includes 25 microfluidic chips, reagents and consumables, sufficient for 300 samples.
Catalog No. NC1762600 Supplier Agilent Technologies Supplier No. 50671506
For the separation, sizing and quantification of dsDNA fragments from 100 to 7500 bp. The kit includes 25 microfluidic chips, reagents and consumables, sufficient for 300 samples.