Learn More
Fluotics Bioassay Dish Qtrays
Catalog No. NC2196012
Encompass
Bioassay Dish Qtrays, sterile, 245 x 245 mm2, Case Size: 20 trays/case, Compare to: X6023
This item is not returnable. View return policy
Catalog No. NC2196012 Supplier Fluotics Supplier No. QTRY48
Bioassay Dish Qtrays, sterile, 245 x 245 mm2, Case Size: 20 trays/case, Compare to: X6023