Learn More
Biochain Institute Inc Urea Assay Kit, 500 assays/PK
SDP
Catalog No. 50179792
Encompass_Preferred
BioChain's urea assay kit is a simple, direct and automation-ready assay for measuring urea concentration or blood urea nitrogen BUN in biological samples without pretreatment. The improved Jung method utilizes a chromogenic reagent that forms a colored complex specifically with urea. The intensity of the color is directly proportional to the urea concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples.
Catalog No. 50-179-792 Supplier Biochain Institute Inc Supplier No. Z5030016
BioChain's urea assay kit is a simple, direct and automation-ready assay for measuring urea concentration or blood urea nitrogen BUN in biological samples without pretreatment. The improved Jung method utilizes a chromogenic reagent that forms a colored complex specifically with urea. The intensity of the color is directly proportional to the urea concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples.