Learn More
Biochain Institute Inc Hemoglobin Assay Kit, 250 Assays/PK
SDP
Catalog No. 50179801
Encompass_Preferred
BioChain's hemoglobin concentration assay kit is a simple, direct and automation-ready assay for measuring hemoglobin. The improved cyanohemiglobin method, in which the hemoglobin is converted into a uniform colored end product. The intensity of the color is directly proportional to the hemoglobin concentration in the sample. The optimized formulation exhibits high sensitivity and substantially reduces interference by substances in the raw samples.
Catalog No. 50-179-801 Supplier Biochain Institute Inc Supplier No. Z5030026
BioChain's hemoglobin concentration assay kit is a simple, direct and automation-ready assay for measuring hemoglobin. The improved cyanohemiglobin method, in which the hemoglobin is converted into a uniform colored end product. The intensity of the color is directly proportional to the hemoglobin concentration in the sample. The optimized formulation exhibits high sensitivity and substantially reduces interference by substances in the raw samples.