Learn More
Biochain Institute Inc Enzyme Ethanol Assay Kit, 100 Assays/PK
SDP
Catalog No. 50179812
Encompass_Preferred
BioChain's ethanol assay kit is a simple, direct and automation-ready assay based on alcohol dehydrogenase catalyzed oxidation of ethanol, in which the formed NADH is coupled to the formazan (MTT) chromogen. The intensity of the product color, measured at 565 nm, is proportionate to the ethanol concentration in the sample.
Catalog No. 50-179-812 Supplier Biochain Institute Inc Supplier No. Z5030040
BioChain's ethanol assay kit is a simple, direct and automation-ready assay based on alcohol dehydrogenase catalyzed oxidation of ethanol, in which the formed NADH is coupled to the formazan (MTT) chromogen. The intensity of the product color, measured at 565 nm, is proportionate to the ethanol concentration in the sample.