Learn More
Biochain Institute Inc Ethanol Assay Kit, 500 Assays/PK
SDP
Catalog No. 50179811
Encompass_Preferred
BioChain's ethanol assay kit is a simple, direct and automation-ready assay based on an improved dichromate method, in which dichromate is reduced by ethanol to a bluish chromic (Cr3+) product. The intensity of color, measured at 580 nm, is a direct measure of the alcohol concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples and exhibits high sensitivity.
Catalog No. 50-179-811 Supplier Biochain Institute Inc Supplier No. Z5030029
BioChain's ethanol assay kit is a simple, direct and automation-ready assay based on an improved dichromate method, in which dichromate is reduced by ethanol to a bluish chromic (Cr3+) product. The intensity of color, measured at 580 nm, is a direct measure of the alcohol concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples and exhibits high sensitivity.