Learn More
Biochain Institute Inc Bradford Protein Assay Kit, 500 Assays/PK
SDP
Catalog No. 50179820
Encompass_Preferred
BioChain's protein assay kit is a simple, direct and automation-ready assay based on an improved Coomassie Blue G method. The dye forms a blue complex specifically with protein, and the intensity of color, measured at 595nm, is directly proportional to the protein concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples and exhibits increased sensitivity towards peptides.
Catalog No. 50-179-820 Supplier Biochain Institute Inc Supplier No. Z5030028
BioChain's protein assay kit is a simple, direct and automation-ready assay based on an improved Coomassie Blue G method. The dye forms a blue complex specifically with protein, and the intensity of color, measured at 595nm, is directly proportional to the protein concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples and exhibits increased sensitivity towards peptides.