Learn More
AMPLYUS LLC miniPCR bio BioBits: Central Dogma (classroom kit)
SDP
Catalog No. NC1896915
Encompass_Preferred
BioBits: Central Dogma. Students observe how genetic information flows from DNA to RNA, and then to protein, right before their eyes — transcription and translation in a test tube, with stunning fluorescent outputs.
Catalog No. NC1896915 Supplier AMPLYUS LLC Supplier No. KT191001
BioBits: Central Dogma. Students observe how genetic information flows from DNA to RNA, and then to protein, right before their eyes — transcription and translation in a test tube, with stunning fluorescent outputs.