Learn More
Cayman Chemical Biochanin A, 491-80-5, 100 mg
SDP
Supplier:  Cayman Chemical 16476100
Encompass_Preferred
A phytoestrogen that can affect hormone levels by inhibiting 5α-reductase and 17β-hydroxysteroid dehydrogenase or altering aromatase (CYP19A1) activity; activates PPARγ (IC50 = 19 µM) and increases the expression of PGC-1α; also inhibits FAAH (IC50 = 2.4 µM) and agonizes AhR (EC50 = 0.25 µM)
Catalog No. NC1344584