Learn More
Selleck Chemical LLC Biochanin A S2377-200mg
SDP
Catalog No. 503109559
Encompass_Preferred
Biochanin A (4-Methylgenistein) an O-methylated isoflavone from Trifolium pratense inhibits protein tyrosine kinase (PTK) of epidermal growth factor receptor with IC50 values of 91 5 M
Catalog No. 50-310-9559 Supplier Selleck Chemical LLC Supplier No. S2377200MG
Biochanin A (4-Methylgenistein) an O-methylated isoflavone from Trifolium pratense inhibits protein tyrosine kinase (PTK) of epidermal growth factor receptor with IC50 values of 91 5 M