Learn More
Apexbio Technology LLC Biochanin A 491-80-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062701
Encompass_Preferred
Biochanin A (CAS number 491-80-5) is a naturally derived inhibitor of fatty acid amide hydrolase (FAAH) an enzyme responsible for metabolizing the endocannabinoid anandamide (AEA) In vitro studies show Biochanin A inhibits FAAH-mediated hydrolysis of AEA with IC50 values of 1 8 M (mouse) 1 4 M (rat) and 2 4 M (human) without affecting CB1 CB2 receptors or FAAH-2 In animal models Biochanin A attenuates formalin-induced phosphorylation of spinal extracellular signal-regulated kinase an effect reversible by CB1 receptor antagonist AM251 Biochanin A thus serves as a useful tool compound in pain regulation and endocannabinoid research
Catalog No. 50-000-62701 Supplier Apexbio Technology LLC Supplier No. N13085.1
Biochanin A (CAS number 491-80-5) is a naturally derived inhibitor of fatty acid amide hydrolase (FAAH) an enzyme responsible for metabolizing the endocannabinoid anandamide (AEA) In vitro studies show Biochanin A inhibits FAAH-mediated hydrolysis of AEA with IC50 values of 1 8 M (mouse) 1 4 M (rat) and 2 4 M (human) without affecting CB1 CB2 receptors or FAAH-2 In animal models Biochanin A attenuates formalin-induced phosphorylation of spinal extracellular signal-regulated kinase an effect reversible by CB1 receptor antagonist AM251 Biochanin A thus serves as a useful tool compound in pain regulation and endocannabinoid research