Learn More
Apexbio Technology LLC Biochanin A, 200mg. Cas: 491-80-5 MFCD: MFCD00006839. Flavonoid isoflavone
SDP
Supplier:  Apexbio Technology LLC N1308200
Encompass_Preferred
IC50: 1.8, 1.4 and 2.4 M for mouse, rat and human FAAH, respectivelyBiochanin A is a natural inhibitor of fatty acid amide hydrolase. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5637
 Restricted Item