Learn More
Medchemexpress LLC Biochanin A | 491-80-5 | 98.7% | C₁₆H₁₂O₅ | 500 MG
SDP
Catalog No. 5000354220
Encompass_Preferred
Biochanin A (Standard) is the analytical standard of Biochanin A, intended for research and analytical applications. It is a naturally occurring fatty acid amide hydrolase (FAAH) inhibitor, demonstrating inhibition of FAAH with IC50s of 1.8, 1.4, and 2.4 μM for mouse, rat, and human FAAH, respectively. This compound serves as an analytical standard and reference standard for assays and is frequently employed in qualitative, quantitative, and methodological research experiments.
  • Inhibits FAAH with IC50s of 1.8, 1.4, and 2.4 μM for mouse, rat, and human FAAH
  • Used as an analytical standard and reference standard for assays
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Compatible with techniques such as HPLC, GC, and MS
Catalog No. 50-003-54220 Supplier Medchemexpress LLC Supplier No. HY14595500MG
Biochanin A (Standard) is the analytical standard of Biochanin A, intended for research and analytical applications. It is a naturally occurring fatty acid amide hydrolase (FAAH) inhibitor, demonstrating inhibition of FAAH with IC50s of 1.8, 1.4, and 2.4 μM for mouse, rat, and human FAAH, respectively. This compound serves as an analytical standard and reference standard for assays and is frequently employed in qualitative, quantitative, and methodological research experiments.
  • Inhibits FAAH with IC50s of 1.8, 1.4, and 2.4 μM for mouse, rat, and human FAAH
  • Used as an analytical standard and reference standard for assays
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Compatible with techniques such as HPLC, GC, and MS