Learn More
Medchemexpress LLC Biochanin A (Standard) | 491-80-5 | 98.7% | C16H12O5 | 25 MG
SDP
Catalog No. 5000352973
Encompass_Preferred
Biochanin A (Standard) is the analytical standard of Biochanin A, intended for research and analytical applications. It is a naturally occurring fatty acid amide hydrolase (FAAH) inhibitor, inhibiting FAAH with IC₅₀s of 1.8, 1.4, and 2.4 μM for mouse, rat, and human FAAH, respectively.
  • Analytical standard of Biochanin A
  • Intended for research and analytical applications
  • Naturally occurring fatty acid amide hydrolase (FAAH) inhibitor
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-52973 Supplier Medchemexpress LLC Supplier No. 25MGHY14595R
Biochanin A (Standard) is the analytical standard of Biochanin A, intended for research and analytical applications. It is a naturally occurring fatty acid amide hydrolase (FAAH) inhibitor, inhibiting FAAH with IC₅₀s of 1.8, 1.4, and 2.4 μM for mouse, rat, and human FAAH, respectively.
  • Analytical standard of Biochanin A
  • Intended for research and analytical applications
  • Naturally occurring fatty acid amide hydrolase (FAAH) inhibitor
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS