Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biochanin A 98.0+%, TCI America™
SDP

Catalog No. B40981G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40981G 1 g
B4098200MG 200 mg
B40985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B40981G Supplier TCI America Supplier No. B40981G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 491-80-5
Molecular Formula C16H12O5
Molecular Weight (g/mol) 284.27
MDL Number MFCD00006839
InChI Key WUADCCWRTIWANL-UHFFFAOYSA-N
Synonym biochanin a, biochanin, 4'-methylgenistein, 5,7-dihydroxy-4'-methoxyisoflavone, 5,7-dihydroxy-3-4-methoxyphenyl-4h-chromen-4-one, genistein 4-methyl ether, pratensol, biochanine a, 5,7-dihydrox-4'-methoxyisoflavone, 4h-1-benzopyran-4-one, 5,7-dihydroxy-3-4-methoxyphenyl
PubChem CID 5280373
ChEBI CHEBI:17574
IUPAC Name 5,7-dihydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one
SMILES COC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O

Specifications

Melting Point 217°C
Quantity 1 g
Formula Weight 284.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Biochanin A
RTECSNumber DJ3002500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.