Learn More
BIOCHAR NOW LLC Biochar Powder, 5 Gallon Bag
SDP
Supplier:  BIOCHAR NOW LLC BIOCHAR POWDER5 GALLON
Encompass
Simply put, biochar is a highly adsorbent, specially-produced charcoal originally used as a soil amendment. Similar to charcoal, biochar is produced using the ancient practice of heating wood or other plant material (biomass) with little to no oxygen. However, unlike charcoal, which is often used for cooking, biochar is made under specific conditions with the intent to be applied to soil as a means to increase soil fertility and agricultural yields, and sequester carbon to reverse global warming. Other market uses of biochar are being discovered regularly as universities and large industrial partners conduct extensive research into this versatile material and its cation exchange properties.
Catalog No. NC1713201