Learn More
Biomatik Corporation T4 DNA Ligase, 5U/uL One pouch containing 2 tubes T4 and 10X buffer. Grade Biotechnology. Storage -20C.
Catalog No. 502741330
Encompass_Preferred
T4 DNA Ligase, 5U/uL One pouch containing 2 tubes T4 and 10X buffer. Grade Biotechnology. Storage -20C.
Catalog No. 50-274-1330 Supplier Biomatik Corporation Supplier No. A1113500U
T4 DNA Ligase, 5U/uL One pouch containing 2 tubes T4 and 10X buffer. Grade Biotechnology. Storage -20C.