Learn More
FOCUS BIOMOLECULES Biochemical Reagents, CDD-2789, ALK1/2 inhibitor, CAS 3052084-77-9, 98% by HPLC/NMR (Conforms)
SDP
Catalog No. NC3802662
Encompass_Preferred
Biochemical Reagents, CDD-2789, ALK1/2 inhibitor, CAS 3052084-77-9, 98% by HPLC/NMR (Conforms), CDD-2789 is a potent (IC50 3.9 nM ALK1/ 2.1 nM ALK2) and selective ALK1/2 inhibitor. It attenuated activin A and BMP-induced phosphorylated SMAD1/5 activation in fibroblasts in a dose-dependent manner.
Catalog No. NC3802662 Supplier FOCUS BIOMOLECULES Supplier No. 10445125MG
Biochemical Reagents, CDD-2789, ALK1/2 inhibitor, CAS 3052084-77-9, 98% by HPLC/NMR (Conforms), CDD-2789 is a potent (IC50 3.9 nM ALK1/ 2.1 nM ALK2) and selective ALK1/2 inhibitor. It attenuated activin A and BMP-induced phosphorylated SMAD1/5 activation in fibroblasts in a dose-dependent manner.