Learn More
FOCUS BIOMOLECULES Biochemical Reagents, DHP-103, Selective KCa3.1 blocker, CAS 2775311-82-3, 98% by HPLC/NMR (Conforms)
SDP
Catalog No. NC3797278
Encompass_Preferred
Biochemical Reagents, DHP-103, Selective KCa3.1 blocker, CAS 2775311-82-3, 98% by HPLC/NMR (Conforms), DHP-103 is a potent (IC50 6 nM) blocker of the intermediate-conductance calcium-activated potassium channel KCa3.1. It displays exquisite selectivity over calcium channels. It blocked mutant KCa3.1 channels that cause hereditary xerocytosis and reduced infarct volume, improved balance beam performance, and reduced activated microglia in a rat model of acute ischemic stroke.
Catalog No. NC3797278 Supplier FOCUS BIOMOLECULES Supplier No. 104906
Add to Cart
Add to Cart
Biochemical Reagents, DHP-103, Selective KCa3.1 blocker, CAS 2775311-82-3, 98% by HPLC/NMR (Conforms), DHP-103 is a potent (IC50 6 nM) blocker of the intermediate-conductance calcium-activated potassium channel KCa3.1. It displays exquisite selectivity over calcium channels. It blocked mutant KCa3.1 channels that cause hereditary xerocytosis and reduced infarct volume, improved balance beam performance, and reduced activated microglia in a rat model of acute ischemic stroke.