Learn More
FOCUS BIOMOLECULES Biochemical Reagents, Dibutyryl-cAMP, Cell permeable cAMP, CAS 16980-89-5, 98% by HPLC/NMR (conforms)
SDP
Catalog No. NC3802458
Encompass_Preferred
Biochemical Reagents, Dibutyryl-cAMP, Cell permeable cAMP, CAS 16980-89-5, 98% by HPLC/NMR (conforms), Cell-permeable cAMP analog which mimics the effect of endogenous cAMP when applied to cells.1 Activates PKA.2,3 Induces morphological differentiation of astrocytes.4 Promotes differentiation of dopaminergic neurons from hPSCs (in cocktails with other agents).5
Catalog No. NC3802458 Supplier FOCUS BIOMOLECULES Supplier No. 102627100MG
Biochemical Reagents, Dibutyryl-cAMP, Cell permeable cAMP, CAS 16980-89-5, 98% by HPLC/NMR (conforms), Cell-permeable cAMP analog which mimics the effect of endogenous cAMP when applied to cells.1 Activates PKA.2,3 Induces morphological differentiation of astrocytes.4 Promotes differentiation of dopaminergic neurons from hPSCs (in cocktails with other agents).5