Learn More
FOCUS BIOMOLECULES Biochemical Reagents, Domoic acid, Marine natural product neurotoxin, CAS 14277-97-5, 98% by HPLC/NMR (Conforms)
SDP
Catalog No. NC3797321
Encompass_Preferred
Biochemical Reagents, Domoic acid, Marine natural product neurotoxin, CAS 14277-97-5, 98% by HPLC/NMR (Conforms), Kainic acid-type neurotoxin produced by certain species of the marine pennate diatom Pseudo-nitzschia, is responsible for amnesic shellfish poisoning.1 Suspension feeders such as bivalve mollusks can accumulate and retain high amounts of DA in their tissues, threatening human health.2,3 DA is a potent agonist at kainate and AMPA receptors4 and induces convulsive behavior and seizures in rodents5. It induces structural and molecular changes in the developing zebra fish nervous system.6
Catalog No. NC3797321 Supplier FOCUS BIOMOLECULES Supplier No. 102472
Biochemical Reagents, Domoic acid, Marine natural product neurotoxin, CAS 14277-97-5, 98% by HPLC/NMR (Conforms), Kainic acid-type neurotoxin produced by certain species of the marine pennate diatom Pseudo-nitzschia, is responsible for amnesic shellfish poisoning.1 Suspension feeders such as bivalve mollusks can accumulate and retain high amounts of DA in their tissues, threatening human health.2,3 DA is a potent agonist at kainate and AMPA receptors4 and induces convulsive behavior and seizures in rodents5. It induces structural and molecular changes in the developing zebra fish nervous system.6