Learn More
FOCUS BIOMOLECULES Biochemical Reagents, Forskolin, Adenylate cyclase activator, CAS 66575-29-9, 98% by HPLC/NMR (Conforms)
Supplier: FOCUS BIOMOLECULES 10207350MG
Biochemical Reagents, Forskolin, Adenylate cyclase activator, CAS 66575-29-9, 98% by HPLC/NMR (Conforms), Forskolin is a widely used adenylate cyclase activator.1 It also is a positive inotropic agent, vasodilator and induces platelet activation among other activities.2-5