Learn More
FOCUS BIOMOLECULES Biochemical Reagents, Gliocidin, Nicotinamide-mimetic prodrug, CAS 62289-81-0, 98% by HPLC/NMR (Conforms)
SDP
Catalog No. NC3796922
Encompass_Preferred
Biochemical Reagents, Gliocidin, Nicotinamide-mimetic prodrug, CAS 62289-81-0, 98% by HPLC/NMR (Conforms), Gliocidin is a nicotinamide-mimetic prodrug that targets de novo purine biosynthesis vulnerability in glioblastoma (IC50 150 nM).1 Gliocidin was converted via the NAD salvage pathway to gliocidin-adenine dinucleotide with directly interfered with the activity of IMPDH2 resulting in guanylate depletion and cell death. Displays strong blood brain barrier permeability and suppression of glioblastoma in athymic nude mice. Gliocidin showed synergistic effects with temozolomide. Inhibitor of the SARS 3CLPRO protease.2
Catalog No. NC3796922 Supplier FOCUS BIOMOLECULES Supplier No. 103952
Biochemical Reagents, Gliocidin, Nicotinamide-mimetic prodrug, CAS 62289-81-0, 98% by HPLC/NMR (Conforms), Gliocidin is a nicotinamide-mimetic prodrug that targets de novo purine biosynthesis vulnerability in glioblastoma (IC50 150 nM).1 Gliocidin was converted via the NAD salvage pathway to gliocidin-adenine dinucleotide with directly interfered with the activity of IMPDH2 resulting in guanylate depletion and cell death. Displays strong blood brain barrier permeability and suppression of glioblastoma in athymic nude mice. Gliocidin showed synergistic effects with temozolomide. Inhibitor of the SARS 3CLPRO protease.2