Learn More
FOCUS BIOMOLECULES Biochemical Reagents, HC-258, Covalent TEAD inhibitor, CAS Unknown, 98% by TLC/NMR (Conforms)
SDP
Catalog No. NC3802512
Encompass_Preferred
Biochemical Reagents, HC-258, Covalent TEAD inhibitor, CAS Unknown, 98% by TLC/NMR (Conforms), HC-258 is a covalent inhibitor of TEAD (transcriptional enhanced associate domain), an important component of the Hippo pathway. It was able to reduce gene expression levels of CTGF, CYR61, AXL, and NF2 in MDA-MB-231 breast cancer cells. HC-258 also reduced MDA-MB-231 cell migration.
Catalog No. NC3802512 Supplier FOCUS BIOMOLECULES Supplier No. 10393125MG
Biochemical Reagents, HC-258, Covalent TEAD inhibitor, CAS Unknown, 98% by TLC/NMR (Conforms), HC-258 is a covalent inhibitor of TEAD (transcriptional enhanced associate domain), an important component of the Hippo pathway. It was able to reduce gene expression levels of CTGF, CYR61, AXL, and NF2 in MDA-MB-231 breast cancer cells. HC-258 also reduced MDA-MB-231 cell migration.