Learn More
FOCUS BIOMOLECULES Biochemical Reagents, Mitomycin C, DNA crosslinking and damaging agent, CAS 50-07-7, 98% by TLC/NMR Conforms
SDP
Catalog No. NC3802453
Encompass_Preferred
Biochemical Reagents, Mitomycin C, DNA crosslinking and damaging agent, CAS 50-07-7, 98% by TLC/NMR Conforms, DNA damaging and crosslinking agent. Induces apoptosis in various cell lines. Cell permeable.This product is not approved for human therapeutic use.
Catalog No. NC3802453 Supplier FOCUS BIOMOLECULES Supplier No. 10117025MG
Biochemical Reagents, Mitomycin C, DNA crosslinking and damaging agent, CAS 50-07-7, 98% by TLC/NMR Conforms, DNA damaging and crosslinking agent. Induces apoptosis in various cell lines. Cell permeable.This product is not approved for human therapeutic use.