Learn More
FOCUS BIOMOLECULES Biochemical Reagents, Mitomycin C, DNA crosslinking and damaging agent, CAS 50-07-7, 98% by TLC/NMR Conforms
SDP
Supplier:  FOCUS BIOMOLECULES 101170
Encompass_Preferred
Biochemical Reagents, Mitomycin C, DNA crosslinking and damaging agent, CAS 50-07-7, 98% by TLC/NMR Conforms, DNA damaging and crosslinking agent. Induces apoptosis in various cell lines. Cell permeable.This product is not approved for human therapeutic use.
Catalog No. NC3796499