Learn More
FOCUS BIOMOLECULES Biochemical Reagents, Oligomycin complex, Mitochondrial ATPase inhibitor, CAS 1404-19-9, Mixture of Oligomycin A (67.5%) B (19.4%) and C (9.8%)/97% by TLC/NMR: (Conforms)
SDP
Catalog No. NC3796505
Encompass_Preferred
Biochemical Reagents, Oligomycin complex, Mitochondrial ATPase inhibitor, CAS 1404-19-9, Mixture of Oligomycin A (67.5%) B (19.4%) and C (9.8%)/97% by TLC/NMR: (Conforms), Inhibits mitochondrial F1F0 ATP synthase1. A useful tool for decreasing cellular ATP levels2. Induces autophagy3. Stimulates lysosome acidification4. Protects against ischemic kidney in male rats5.
Catalog No. NC3796505 Supplier FOCUS BIOMOLECULES Supplier No. 102092
Biochemical Reagents, Oligomycin complex, Mitochondrial ATPase inhibitor, CAS 1404-19-9, Mixture of Oligomycin A (67.5%) B (19.4%) and C (9.8%)/97% by TLC/NMR: (Conforms), Inhibits mitochondrial F1F0 ATP synthase1. A useful tool for decreasing cellular ATP levels2. Induces autophagy3. Stimulates lysosome acidification4. Protects against ischemic kidney in male rats5.