Learn More
FOCUS BIOMOLECULES Biochemical Reagents, STING Degrader 2, STING molecular glue, CAS Unknown, 98% by HPLC/NMR (Conforms)
SDP
Supplier:  FOCUS BIOMOLECULES 103939
Encompass_Preferred
Biochemical Reagents, STING Degrader 2, STING molecular glue, CAS Unknown, 98% by HPLC/NMR (Conforms), STING Degrader 2 is a molecular glue that covalently binds both STING and RNF126 E3 ligase leading to proteasomal degradation. It displayed sustained STING degradation with faster onset and at lower concentration (3 uM) when compared to the STING PROTAC SP23.
Catalog No. NC3796667