Learn More
FOCUS BIOMOLECULES Biochemical Reagents, T-10418, G2A agonist, CAS 1608784-68-4, 98% by HPLC/NMR (Conforms)
SDP
Catalog No. NC3797288
Encompass_Preferred
Biochemical Reagents, T-10418, G2A agonist, CAS 1608784-68-4, 98% by HPLC/NMR (Conforms), T-10418 is a potent (EC50 820 nM) and selective (against 300 non-olfactory GPCRs) agonist of G2 accumulation protein (G2A/ GPR132). At 100 nM, T-10418 resulted in 244% TRPV1 sensitization in mouse sensory neurons. T-10418 is an interesting new chemical tool to study G2A biology.
Catalog No. NC3797288 Supplier FOCUS BIOMOLECULES Supplier No. 104916
Biochemical Reagents, T-10418, G2A agonist, CAS 1608784-68-4, 98% by HPLC/NMR (Conforms), T-10418 is a potent (EC50 820 nM) and selective (against 300 non-olfactory GPCRs) agonist of G2 accumulation protein (G2A/ GPR132). At 100 nM, T-10418 resulted in 244% TRPV1 sensitization in mouse sensory neurons. T-10418 is an interesting new chemical tool to study G2A biology.