Learn More
HARVARD BIOSCIENCE INC Biochrom Ninhydrin Reagent, Amino Acid Analysis, Liquid, 2 L
Catalog No. NC3955210
Encompass
This Ultra Ninhydrin Kit is an analyte specific reagent designed to produce a color reaction with primary and secondary amino acids as well as certain amines. It is intended for use as a component in laboratory-developed tests for amino acid analysis via ion exchange liquid chromatography and post-column derivatization. The kit is supplied in a 2L format and is formulated for compatibility with dedicated amino acid analyzer systems.
  • Designed for colorimetric detection of primary and secondary amino acids
  • Compatible with post-column derivatization methods
  • Supplied as a 2L volume
  • Stable under normal conditions when protected from direct light
  • Intended for professional laboratory use
Catalog No. NC3955210 Supplier HARVARD BIOSCIENCE INC Supplier No. 80600310
This Ultra Ninhydrin Kit is an analyte specific reagent designed to produce a color reaction with primary and secondary amino acids as well as certain amines. It is intended for use as a component in laboratory-developed tests for amino acid analysis via ion exchange liquid chromatography and post-column derivatization. The kit is supplied in a 2L format and is formulated for compatibility with dedicated amino acid analyzer systems.
  • Designed for colorimetric detection of primary and secondary amino acids
  • Compatible with post-column derivatization methods
  • Supplied as a 2L volume
  • Stable under normal conditions when protected from direct light
  • Intended for professional laboratory use