Learn More
Axion Biosystems BIOCIRCUIT MEA 96
SDP
Catalog No. NC2299954
Encompass
The Biocircuit MEA plates deliver high quality results together with industry-leading throughput at the lowest cost per well. 96 well, electrodes wells 8 gold, bottom opaque, walls clear.AXION BIOSYSTEMS mfg# M768-BIO-96
Catalog No. NC2299954 Supplier Axion Biosystems Supplier No. M768BIO96
The Biocircuit MEA plates deliver high quality results together with industry-leading throughput at the lowest cost per well. 96 well, electrodes wells 8 gold, bottom opaque, walls clear.AXION BIOSYSTEMS mfg# M768-BIO-96