Learn More
Azenta US Inc BIOCISION CRYOBOXES - ORANGE
Catalog No. NC2294471
Encompass_Preferred
BioCision® TruCool™ Cryoboxes - Orange. 81 hole. 133L×133W×51D mm (5.25×5.25×2"). Pack of 5. BCS-206O
This item is not returnable. View return policy
Catalog No. NC2294471 Supplier Azenta US Inc Supplier No. BCS206O
BioCision® TruCool™ Cryoboxes - Orange. 81 hole. 133L×133W×51D mm (5.25×5.25×2"). Pack of 5. BCS-206O