Learn More
Research Products International Corp BioClave, 8 Digital
SDP
Catalog No. 501976815
Encompass
BioClave Digital Benchtop Autoclaves are designed for steam sterilization of laboratory instruments and consumables at an economical price. Compact exterior size allows either model to fit comfortably on even the most crowded benchtop. Stainless steel chambers are large enough to accommodate a variety of vessels, glassware, plasticware and other utensils. Features include automatic operation, mechanical and electrical safety interlocks; stainless steel trays, tray removal tool and large digital display.
BioClave 8 has an 8 liter capacity and preset to meet basic sterilization requirements. Temperature can be selected at either 120°C or 134°C with a fixed sterilization time.
Two year warranty.
Catalog No. 50-197-6815 Supplier Research Products International Corp Supplier No. 248558
Add to Cart
Add to Cart
Request a Quote
BioClave Digital Benchtop Autoclaves are designed for steam sterilization of laboratory instruments and consumables at an economical price. Compact exterior size allows either model to fit comfortably on even the most crowded benchtop. Stainless steel chambers are large enough to accommodate a variety of vessels, glassware, plasticware and other utensils. Features include automatic operation, mechanical and electrical safety interlocks; stainless steel trays, tray removal tool and large digital display.
BioClave 8 has an 8 liter capacity and preset to meet basic sterilization requirements. Temperature can be selected at either 120°C or 134°C with a fixed sterilization time.
Two year warranty.