Learn More
Chemglass Life Sciences BioClave 8L Benchtop Autoclave Mini, 120V
SDP
Catalog No. 50143352
Encompass
BioClave 8L Benchtop Autoclave Mini, 120V. With a width of less than 14¿ for the BioClave Mini, this model will fit comfortably on even the most crowded bench tops.Despite the space-saving exterior dimensions, the sterilization chambers are large, accommodating a variety of liquids, media, instruments, glassware, plastic ware and other common laboratory items. This model includes a mechanical and electrical safety interlock that prevents the door from being opened until the pressure has reached zero PSI. Two trays are supplied with each Benchtop BioClave Mini, along with a convenient tray removal tool.The 8 liter BioClave Mini is preprogrammed to meet basic sterilization needs. Although the temperature can be set at either 121¿C or 134¿C, the sterilization time is fixed. The economical BioClave Mini is the ideal choice wherever bench top sterilization is required and space is limited.
Catalog No. 50-143-352 Supplier Chemglass Life Sciences Supplier No. CLS1191008
Add to Cart
Add to Cart
Request a Quote
BioClave 8L Benchtop Autoclave Mini, 120V. With a width of less than 14¿ for the BioClave Mini, this model will fit comfortably on even the most crowded bench tops.Despite the space-saving exterior dimensions, the sterilization chambers are large, accommodating a variety of liquids, media, instruments, glassware, plastic ware and other common laboratory items. This model includes a mechanical and electrical safety interlock that prevents the door from being opened until the pressure has reached zero PSI. Two trays are supplied with each Benchtop BioClave Mini, along with a convenient tray removal tool.The 8 liter BioClave Mini is preprogrammed to meet basic sterilization needs. Although the temperature can be set at either 121¿C or 134¿C, the sterilization time is fixed. The economical BioClave Mini is the ideal choice wherever bench top sterilization is required and space is limited.