Learn More
Alkali Scientific BioCleanC™ Chemotherapy Protective Apron with Sleeves, Size Small, 40 /cs
SDP
Catalog No. 502388764
Encompass_Preferred
Providing protection against a range of chemotherapy drugs, the BioClean-C™ Chemotherapy Protective Apron with Sleeves is manufactured from lightweight, low-linting CleanTough™ blue material, and features tie tapes at the rear and an adjustable neck fastening for easy donning and comfort. Tested against permeation standard ASTM F739-12.
Catalog No. 50-238-8764 Supplier Alkali Scientific Supplier No. A931
Providing protection against a range of chemotherapy drugs, the BioClean-C™ Chemotherapy Protective Apron with Sleeves is manufactured from lightweight, low-linting CleanTough™ blue material, and features tie tapes at the rear and an adjustable neck fastening for easy donning and comfort. Tested against permeation standard ASTM F739-12.