Learn More
Alkali Scientific BioCleanC™ Chemotherapy Protective Apron, Size Medium, 50/cs
SDP
Catalog No. 502388816
Encompass_Preferred
Providing protection against a range of chemotherapy drugs, the BioClean-C™ Chemotherapy Protective Apron is manufactured from lightweight, low-linting CleanTough™ blue material, and features tie tapes at the rear and an adjustable neck fastening for easy donning and comfort. Tested against permeation standard ASTM F739-12.
Catalog No. 50-238-8816 Supplier Alkali Scientific Supplier No. A922
Providing protection against a range of chemotherapy drugs, the BioClean-C™ Chemotherapy Protective Apron is manufactured from lightweight, low-linting CleanTough™ blue material, and features tie tapes at the rear and an adjustable neck fastening for easy donning and comfort. Tested against permeation standard ASTM F739-12.