Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask
GREENER_CHOICE

Catalog No. 0877436 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
250 mL
750 mL
Culture Area:
175 cm2
75 cm2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Culture Area
08-774-36 750 mL 175 cm2
08-774-35 250 mL 75 cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08-774-36 Supplier Corning™ Supplier No. 354528
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology. Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask is ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.

 

  • BioCoat™ provides a superior solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures
  • Matrix components are applied as an even, optically clear coating
  • All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells

 

For research use only.

TRUSTED_SUSTAINABILITY

Specifications

Closure Material Phenolic
Color Clear
Capacity (Metric) 750 mL
Material Polystyrene
For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
Neck Style Canted
Product Type Cell Culture Flask
Shape Rectangular
Surface Coating Collagen IV
Sterility Sterile
Bottom Shape Flat
Closure Type Plug Seal
Culture Area 175 cm2
Volume (Metric) Working 35 to 52.5 mL
Quantity 5 Ea.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.