Learn More
Description
Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces, simulating the natural environment of cells in vivo.
- Source: Mouse
- TC-treated
- Nonsterile
For adhesion, growth and differentiation of neuronal and transfected cell lines
Specifications
Specifications
| Bottom Shape | Round |
| Color | Clear |
| Culture Area | 21.3 cm2 |
| Material | Polystyrene |
| Shape | Round |
| Surface Treatment | Collagen Type IV |
| Type | Cell Culture Dish |
| No. per Case | 20/Cs. |
| No. per Pack | 5/Pk. |
| Diameter (Metric) | 60 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
