Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ Laminin Rectangular Canted Neck Cell Culture Flask w/Plug Seal Cap
GREENER_CHOICE

Catalog No. 0877491 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
250 mL
Culture Area:
75 cm2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Culture Area
08-774-91 250 mL 75 cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-774-91 Supplier Corning™ Supplier No. 354522
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tested for their ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse and human neuroblastoma cells

  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
  • Matrix components are applied as an even, optically clear coating
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells

These products are for research use only and are not intended for diagnostic purposes.

For adhesion, growth and differentiation of neuronal and transfected cell lines

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 8.45 oz.
Closure Material Phenolic
Capacity (Metric) 250 mL
Material Polystyrene
For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
Neck Style Canted
No. per Case 10/Cs.
No. per Pack 5/Pk.
Shape Rectangular
Surface Coating Laminin
Sterility Sterile
Bottom Shape Rectangular
Culture Area 75 cm2
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.