Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ Poly-D-Lysine Vented Cap Flasks
GREENER_CHOICE

Catalog No. 08774334 Shop All Corning Products
Change view
Click to view available options
Culture Area:
150 cm2
175 cm2
25 cm2
75 cm2
No. per Case:
10/cs.
40/cs.
5/cs.
50/cs.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area No. per Case
08-774-334 150 cm2 5/cs.
08-774-333 75 cm2 5/cs.
08-774-335 175 cm2 5/cs.
08-772-60 75 cm2 50/cs.
08-774-344 175 cm2 40/cs.
08-774-341 25 cm2 50/cs.
08-774-343 150 cm2 40/cs.
08-774-332 25 cm2 10/cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 08-774-334 Supplier Corning™ Supplier No. 354538
Only null left
Add to Cart
Add to Cart

Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines

Corning™ BioCoat™ products come in a variety of pre-coated options and are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 20.2 oz.
Closure Material Phenolic
Capacity (Metric) 600 mL
For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
Neck Style Canted
No. per Case 5/cs.
Shape Rectangular
Surface Coating Poly-D-Lysine
Sterility Sterile
Culture Area 150 cm2

Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.