Learn More
AURORA PRO SCIENTIFIC BIOCOMMA EMPTY FPLC COLUMNS5ML
Catalog No. NC3918898
Encompass
Biocomma Empty FPLC Columns with Threaded End, Capacity 5 mL with Green Sealing Sleeve, Super Pressure Resistance
Catalog No. NC3918898 Supplier AURORA PRO SCIENTIFIC Supplier No. MPPC0058
Biocomma Empty FPLC Columns with Threaded End, Capacity 5 mL with Green Sealing Sleeve, Super Pressure Resistance