Learn More
Research Products International Corp Biocooler 2 Bench-Top Cooler, 2.0ml Cryogenic Tubes, Holds 42
SDP
Catalog No. 501977392
Encompass
Biocooler is designed to consistently maintain subzero temperatures of micro-tubes. Benchtop freezer keeps enzymes and valuable samples at -20°C to -15°C for up to 6 hours at room temperature. Protects samples from temperature fluctuation due to frost-free cycles, power loss, or open freezer doors. Features inner note panel for referencing samples and carrying handle for convenient transporting.
Catalog No. 50-197-7392 Supplier Research Products International Corp Supplier No. 260101
Biocooler is designed to consistently maintain subzero temperatures of micro-tubes. Benchtop freezer keeps enzymes and valuable samples at -20°C to -15°C for up to 6 hours at room temperature. Protects samples from temperature fluctuation due to frost-free cycles, power loss, or open freezer doors. Features inner note panel for referencing samples and carrying handle for convenient transporting.