Learn More
NANOMICRO TECHNOLOGIES INC NanoChrom Technologies, BioCore HIC-OH LC Analytical Column, 5 m, 4.6100 mm, Stainless Steel
Catalog No. NC3587964
Encompass_Preferred
BioCore HIC is a family of high-performance, hydrophobic interaction chromatography columns designed for Drug-Antibody-Ratio (DAR) determination in Antibody-Drug Conjugates (ADCs) based on the difference in surface hydrophobicity. The column technology involves the creation of a hydrophilic layer decorated with selective hydrophobic functionality, on the surface of monodispersed, wide-pore, spherical, silica particles, combined with well-established column packing processes.
Catalog No. NC3587964 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B75305010004610S
BioCore HIC is a family of high-performance, hydrophobic interaction chromatography columns designed for Drug-Antibody-Ratio (DAR) determination in Antibody-Drug Conjugates (ADCs) based on the difference in surface hydrophobicity. The column technology involves the creation of a hydrophilic layer decorated with selective hydrophobic functionality, on the surface of monodispersed, wide-pore, spherical, silica particles, combined with well-established column packing processes.