Learn More
NANOMICRO TECHNOLOGIES INC NanoChrom Technologies, BioCore RP-1000 Analytical Column, 5 m, 10150 mm, Stainless Steel
Catalog No. NC3587975
Encompass_Preferred
BioCore RP-1000 is a family of high-performance, reversed-phase columns based on monodispersed, wide-pore, PS/DVB particles. They are designed for separation and determination of exact mass of variants in proteins and their fragments, such as mAbs and related substances.
Catalog No. NC3587975 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. B81105010010015S
BioCore RP-1000 is a family of high-performance, reversed-phase columns based on monodispersed, wide-pore, PS/DVB particles. They are designed for separation and determination of exact mass of variants in proteins and their fragments, such as mAbs and related substances.